Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium iodide solution 30 g/L

Potassium iodide solution 30 g/L

100 mL
Catalog # HACH34332
Supplier:   Hach
Potassium iodide solution 30 g/L
Catalog # HACH34332
Supplier:  Hach
CAS Number:  7681-11-0
Potassium iodide solution 30 g/L
Potassium iodide solution 30 g/L

100 mL

CAS Number:  7681-11-0
Price
23,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Reagent for use in iodometric titrations.

Method: 8210

Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.