Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Hardness 2 indicator solution, 100 ml MDB
Hardness 2 indicator solution, 100 ml MDB
Hardness 2 indicator solution, 100 ml MDB
Catalog # HACH42532
Supplier:
Hach
CAS Number:
Expand image
Hardness 2 indicator solution, 100 ml MDB
Catalog # HACH42532
Supplier:
Hach
CAS Number:
Specifications
Pk:
100 mL
Description:
Hardness 2 indicator solution, 100 mL MDB
Specifications
About this item
Product Details
Product Details