Orders LinkContactUsLinkComponent
pH indicator strips
pH indicator strips
pH indicator strips, 0 - 14,0, 1
Catalog # HACH2601300
Supplier:   Hach
pH indicator strips
Catalog # HACH2601300
Supplier:  Hach
pH indicator strips
pH indicator strips
pH indicator strips, 0 - 14,0, 1
Price
54,30 €
Log in for availability
1 * 100 ST
Product Details & Documents
pH strips provides a simple, fast and economical method of pH measurement. Can obtain accurate results in weakly buffered, turbid and lightly coloured samples. It is easy to use and disposable.
Specifications
  • pH gradation:
    1
  • pH range:
    0 - 14,0
  • Description:
    pH indicator strips