Order Entry
Germany
Orders LinkContactUsLinkComponent
Visual colour kits, phosphate

Visual colour kits, phosphate

Total phosphate test kit, model PO-24, 0 - 0,8 mg/l
0 - 4 mg/l
0 - 40 mg/l, 1 KIT
Catalog # HACH225001
Supplier:   Hach
Visual colour kits, phosphate
Catalog # HACH225001
Supplier:  Hach
CAS Number:  
Visual colour kits, phosphate
Visual colour kits, phosphate

Total phosphate test kit, model PO-24, 0 - 0,8 mg/l
0 - 4 mg/l
0 - 40 mg/l, 1 KIT

CAS Number:  
Price
457,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

This kit provides measurement of ortho- (reactive) and meta- (condensed) phosphate (requires hot acid digestion).

Delivery information: Kit includes five reagents, colour disc, comparator box, viewing adapter, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, funnel, filter paper, deionized water, instructions and carrying case.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Total phosphate test kit, model PO-24
  • Range:
    0 - 0,8 mg/l<br />0 - 4 mg/l<br />0 - 40 mg/l

Specifications

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Colour viewing tube, glass
    Colour viewing tube, glass
    HACH173006
    6
    Log in for availability
    1 * 6 ST - 47,80 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Stoppers for glass viewing tube
    Stoppers for glass viewing tube
    HACH173106
    6
    Log in for availability
    1 * 6 ST - 19,50 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Colour comparator box
    Colour comparator box
    HACH173200
    1
    Log in for availability
    1 * 1 ST - 23,20 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Colour viewing tube, plastic with cap
    Colour viewing tube, plastic with cap
    HACH4660004
    4
    Log in for availability
    1 * 4 ST - 32,40 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Case kit, blue PP, 220×170×150 mm
    Case kit, blue PP, 220×170×150 mm
    HACH4660200
    1
    Log in for availability
    1 * 1 ST - 34,40 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.