Kjeldahl tablets
: Merck
In addition to the traditional 5 g Kjeldahl tablets, which are added as catalysts to the digestion mixture, Merck Millipore can offer the 3,5 g tablet. This gives the flexibility needed to find the optimal setting for the catalyst/sulphuric acid ratio.
- The 3,5 g tablets are suitable for microdigestion
- Easy to use PE packaging concept