ContactUsLinkComponent
Caps for culture tubes
Caps for culture tubes
Catalog # 705-1201
Supplier:  Hach
undefined
Caps for culture tubes
Catalog # 705-1201
Supplier:  Hach
Specifications
  • Thread:
    15-415
  • Colour:
    White
  • Description:
    Screw cap PP with fluoropolymer liner
  • Pk:
    6
Specifications
About this item
Screw cap with fluoropolymer liner.
  • Firmly bonded liner provides a highly inert sealing surface