The Proline Edition X range of heating thermostats feature a wide temperature range, high cooling and heating power and a multi level pump. The self-explanatory menu-guided remote control Command is particularly convenient for operating the units at distances as far as 50 meters away. Thanks to the large display, the user can keep an eye on all the important parameters. There is a graphical display of measured values, large choice of interfaces (e.g. Ether- CAT, Ethernet, RS232/RS485, analogue module, Profibus, contact modules). Data collection together with documentation and control is easy using the Wintherm Plus software. Units are ideal for temperature control of chemical reactors, distillation plants and glass condensers, crystallisation, thermostating measurement setups in process technology, chemical probes, calibration of sensors and testing electronic components.
- Easy to use with simple, intuitive control
- PowerAdapt system provides maximum heating performance, up to 3,5 kW depending on the network in use
- SmartCool system gives to 75% energy savings thanks to digital refrigeration management
- Powerful Varioflex pump (pressure-suction pump) with eight pump levels (except model P12 C)
- Flexible system with various interface modules, lateral and rear pump connections
- Multifunctional: Simultaneous connection of two external applications is possible
Max. flow rate / max. pressure:
25 L/min pressure and 23 L/min suction (except P 12 C: 32 L/min pressure only)
0,7 bar pump pressure and 0,4 bar suction performance (except P 12 C: 1.1 bar pressure only)