Surfactant, RapiGest™ SF
Catalog # WATE186001861
Supplier: WATERS
CAS Number:
Specifications
- Pk:5
- Description:Anionic surfactant, 1 mg, for protein sample preparation in mass spectrometry
Specifications
About this item
RapiGest™ SF is an anionic surfactant for peptide digestion. Supplied as five 1 mg vials for 1 to 50 digestions. The digestion speed and more peptide recovery is enhanced by using trypsin or other enzymes.