Split/splitless inlet seals
Supplier: Agilent
Certificates
About this item
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Delivery: An inlet washer is provided with each inlet seal.