Accessories for turbidity sensor VisoTurb® 900-P
: WTW
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
pH buffers, ready to use
: WTW