Culture flasks with baffles
Supplier: LAT - Labor- & Analysen-Technik GmbH
Certificates
About this item
Culture flasks, made of borosilicate glass, for strong turbulence when stirring in Erlenmeyer flasks.
- For Kapsenberg caps
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Frequently Bought Together