Cetrimonium bromide ≥98.0%, BAKER ANALYZED®, J.T.Baker®
Catalog # N121-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass, amber, wide mouth
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (C₁₉H₄₂BrN):>= 98.0 %
- Insoluble Matter:<= 0.005 %
- Residue after Ignition:<= 0.10 %
- Appearance:Passes Test
- Identification (by IR):Passes Test
Specification Test Results
About this item
:
:364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
Flash Pt:244 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-311-3
UN:3077
ADR:9,III
:GHS07
:GHS09
:GHS05
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772