
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8991-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:99.4 - 100.6 %
- For Laboratory,Research,or Manufacturing Use:99.4 - 100.6 %
- ACS - Assay (C₁₀H₁₆N₂O₈):99.4 - 100.6 %
- ACS - Insoluble in Dilute NH₄OH:≤0.005 %
- ACS - Residue after Ignition:≤0.2 %
- ACS - Nitrilotriacetic Acid:≤0.1 %
- ACS - Calcium (Ca):≤0.001 %
- ACS - Heavy Metals (as Pb):≤0.001 %
- ACS - Iron (Fe):≤0.005 %
- Trace Impurities - Magnesium (Mg):≤5 ppm
Specification Test Results
About this item
:
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541