Tubing connectors
POPR6169IND
- Description:Male luer lock to hose end for 1/4" to 5/16" I.D. tubing
- Seq:04
- Pk:1
These tubing connectors are used in hospitals, laboratories, and biomedical instruments. The metal connectors are precision machined and are made from either nickel plated brass or 316 stainless steel.
Standard luer tapers meets ISO 594 specifications include Female Luer Lock (FLL), Micro-Mate® Female Luer Lock (MM FLL), Male Luer Lock (MLL) and Male Luer (ML).