Orders LinkContactUsLinkComponent
Test panels
Test panels
Supplier:   Industrial Physics
Test panels
Supplier:  Industrial Physics
Certificates
Test panels
22 Options Available Below
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
  • Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
  • Physical testing: Adhesion, elongation, impact resistance and elasticity
  • Appearance testing of coatings (gloss, colour, texture)
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • SIMEVF8522
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    50×100×0,8 mm
    Log in for availability
    Log in for availability
    1 * 1.200 ST
    1 * 1.200 ST - 1 050,00 €
    1 050,00 €
    1 * 1.200 ST
       1 050,00 €
  • SIMEVF8530
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    60×140×0,6 mm
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 560,00 €
    560,00 €
    1 * 600 ST
       560,00 €
  • SIMEVF8533
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    60×140×0,6 mm
    Log in for availability
    Log in for availability
    1 * 900 ST
    1 * 900 ST - 647,00 €
    647,00 €
    1 * 900 ST
       647,00 €
  • SIMEVF8523
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 568,00 €
    568,00 €
    1 * 500 ST
       568,00 €
  • SIMEVF8527
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    75×200×0,8 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 442,00 €
    442,00 €
    1 * 300 ST
       442,00 €
  • SIMEVF8529
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    100×200×0,6 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 517,00 €
    517,00 €
    1 * 400 ST
       517,00 €
  • SIMEVF8521
    Test panel, aluminium, matt, degreased and cleaned
    Test panel, aluminium, matt, degreased and cleaned
    200×300×0,6 mm
    Log in for availability
    Log in for availability
    1 * 150 ST
    1 * 150 ST - 829,00 €
    829,00 €
    1 * 150 ST
       829,00 €
  • SIMEVF8531
    Test panel, aluminium, matt, non-degreased
    Test panel, aluminium, matt, non-degreased
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 445,00 €
    445,00 €
    1 * 500 ST
       445,00 €
  • SIMEVF8532
    Test panel, aluminium, matt, non-degreased
    Test panel, aluminium, matt, non-degreased
    75×150×1,0 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 460,00 €
    460,00 €
    1 * 400 ST
       460,00 €
  • SIMEVF8518
    Test panel, aluminium, chessboard printed, degreased and cleaned
    Test panel, aluminium, chessboard printed, degreased and cleaned
    100×300×0,5 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 2 720,00 €
    2 720,00 €
    1 * 300 ST
       2 720,00 €
  • SIMEVF8524
    Test panel, aluminium, matt, chromated alodine
    Test panel, aluminium, matt, chromated alodine
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 020,00 €
    1 020,00 €
    1 * 500 ST
       1 020,00 €
  • SIMEVF8525
    Test panel, aluminium, matt, chromated alodine
    Test panel, aluminium, matt, chromated alodine
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 020,00 €
    1 020,00 €
    1 * 500 ST
       1 020,00 €
  • SIMEVF8526
    Test panel, aluminium, matt, chromo-free
    Test panel, aluminium, matt, chromo-free
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 070,00 €
    1 070,00 €
    1 * 500 ST
       1 070,00 €
  • SIMEVF8520
    Test panel, aluminium, matt, PVC coated one side
    Test panel, aluminium, matt, PVC coated one side
    200×300×0,6 mm
    Log in for availability
    Log in for availability
    1 * 150 ST
    1 * 150 ST - 450,00 €
    450,00 €
    1 * 150 ST
       450,00 €
  • SIMEVF8519
    Test panel, aluminium, mirror polished, PVC coated one side
    Test panel, aluminium, mirror polished, PVC coated one side
    60×140×0,5 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 692,00 €
    692,00 €
    1 * 500 ST
       692,00 €
  • SIMEVF8510
    Test panel, cold rolled steel, smooth, degreased and cleaned
    Test panel, cold rolled steel, smooth, degreased and cleaned
    60×200×0,5 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 303,00 €
    303,00 €
    1 * 400 ST
       303,00 €
  • SIMEVF8511
    Test panel, cold rolled steel, smooth, degreased and cleaned
    Test panel, cold rolled steel, smooth, degreased and cleaned
    75×150×0,5 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 329,00 €
    329,00 €
    1 * 400 ST
       329,00 €
  • SIMEVF8512
    Test panel, cold rolled steel, smooth, degreased and cleaned
    Test panel, cold rolled steel, smooth, degreased and cleaned
    100×200×0,5 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 363,00 €
    363,00 €
    1 * 250 ST
       363,00 €
  • SIMEVF8513
    Test panel, cold rolled steel, matt, degreased and cleaned
    Test panel, cold rolled steel, matt, degreased and cleaned
    75×150×0,8 mm
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 305,00 €
    305,00 €
    1 * 240 ST
       305,00 €
  • SIMEVF8516
    Test panel, harmonic steel, smooth, degreased and cleaned
    Test panel, harmonic steel, smooth, degreased and cleaned
    60×140×0,5 mm
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 525,00 €
    525,00 €
    1 * 600 ST
       525,00 €
  • SIMEVF8515
    Test panel, harmonic steel, smooth, degreased and cleaned
    Test panel, harmonic steel, smooth, degreased and cleaned
    120×200×0,15 mm
    Log in for availability
    Log in for availability
    1 * 700 ST
    1 * 700 ST - 764,00 €
    764,00 €
    1 * 700 ST
       764,00 €
  • SIMEVF8517
    Test panel, harmonic steel, smooth, degreased and cleaned
    Test panel, harmonic steel, smooth, degreased and cleaned
    150×250×0,15 mm
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 597,00 €
    597,00 €
    1 * 450 ST
       597,00 €