Orders LinkContactUsLinkComponent
Mob caps, PP
Mob caps, PP
Mob caps, PP
Supplier:  VWR Collection
Certificates
Mob caps, PP
8 Options Available Below
Product Details & Documents
Disposable mob caps made of PP. Ideal for use in non critical environments.
  • With single latex-free elastic band
  • Available in different colours
Weight: 10 to 12 g/m²
Packaging: 100 pieces per polybag bag and 1000 pieces per carton.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-9194
  • 113-9195
  • 113-9196
  • 113-9197
  • 113-9199
  • 113-9200
  • 113-9203
  • 113-9204
  • Garment application:
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
Specifications
Product Family Options
Product Information
  • Colour
    Ø
    Availability
    Price
  • White
    520 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST
    Specifications:
    More
    Specifications
    Cat. No.
    113-9194
    Garment application
    General Purpose
  • Blue
    520 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST
    Specifications:
    More
    Specifications
    Cat. No.
    113-9195
    Garment application
    General Purpose
  • Green
    520 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST
    Specifications:
    More
    Specifications
    Cat. No.
    113-9196
    Garment application
    General Purpose
  • Red
    520 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST
    Specifications:
    More
    Specifications
    Cat. No.
    113-9197
    Garment application
    General Purpose
  • White
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 35,90 €
    35,90 €
    1 * 1.000 ST
       35,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-9199
    Garment application
    General Purpose
  • Blue
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 36,30 €
    36,30 €
    1 * 1.000 ST
       36,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-9200
    Garment application
    General Purpose
  • Yellow
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 36,30 €
    36,30 €
    1 * 1.000 ST
       36,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-9203
    Garment application
    General Purpose
  • Purple
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 30,60 €
    30,60 €
    1 * 1.000 ST
       30,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-9204
    Garment application
    General Purpose