Chemical resistant gloves, AlphaTec®
Supplier: Ansell
Certificates
About this item
These cotton jersey gauntlet style gloves with top quality neoprene coating provide protection against mechanical hazards and against a wide range of oils, acids, caustics, alcohols and many solvents.
- Special Hi-Lo insulated gauntlet option with quilted cotton liner, for use in both hot and cold temperatures
- Permits intermittent handling in hot liquids up to 180 °C and handling in cold temperatures down to −25 °C
- Comfortable anatomical glove shape improves dexterity and reduces hand fatigue
Ideal for people working with petrochemicals, oils and chemicals, especially caustics and solvents. Also suitable for use in metal fabrication, refining and plating.
Colour: Black
EN 374 (AKL), EN 388 (3121); Cat. III; AQL 1,5
Caution: Do not use glove 09-022 in applications above 180 °C.
Certifications: Cat. III; AQL 1,5.
Caution: Do not use glove 09-022 in applications above 180 °C.
Specifications
- Catalog No:
- 112-3315
- ANSE09-922/10
- 112-0268
- 112-6186
- ANSE09-928/10
- Model:
- 09-924
- 09-922
- 09-430
- 09-022
- 09-928
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B), EN 407 (X1XXXX)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B), EN 407 (X1XXXX)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B), EN 407 (X1XXXX)
- EN ISO 374-1 Type A (AJKLPT), EN ISO 374-5, EN 388:2016 (3232C), EN 407 (32XXXX), EN 511 (111)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B), EN 407 (X1XXXX)