

Acetic acid glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®
Catalog # 9513-07
Supplier: Avantor
CAS Number:
Specifications
- Pk:19 L
- Seq:5282
Specifications
Specification Test Results
- Assay (CH₃COOH) (by freezing point):≥ 99.7 %
- Color (APHA):≤ 10
- Acetic Anhydride ((CH₃CO)₂O):≤ 0.01 %
- Acetaldehyde:≤ 0.05 %
- Residue after Evaporation:≤ 4 ppm
- Solubility in H₂O:Passes Test
- Specific Gravity at 20°/20°C:≥ 1.048
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤ 1 ppm
- Phosphate (PO₄):≤ 1 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Arsenic and Antimony (as As):≤ 5.0 ppb
- Trace Impurities - Barium (Ba):≤ 10.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 50 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 10.0 ppb
- Trace Impurities - Calcium (Ca):≤ 200.0 ppb
- Trace Impurities - Chromium (Cr):≤ 30.0 ppb
- Trace Impurities - Cobalt (Co):≤ 10.0 ppb
- Trace Impurities - Copper (Cu):≤ 20.0 ppb
- Trace Impurities - Gallium (Ga):≤ 10.0 ppb
- Trace Impurities - Germanium (Ge):≤ 10.0 ppb
- Trace Impurities - Gold (Au):≤ 10.0 ppb
- Heavy Metals (as Pb):≤ 300.0 ppb
- Trace Impurities - Iron (Fe):≤ 100.0 ppb
- Trace Impurities - Lead (Pb):≤ 100 ppb
- Trace Impurities - Lithium (Li):≤ 10.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 50.0 ppb
- Trace Impurities - Manganese (Mn):≤ 10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 10.0 ppb
- Trace Impurities - Nickel (Ni):≤ 25.0 ppb
- Trace Impurities - Niobium (Nb):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 50 ppb
- Trace Impurities - Silver (Ag):≤ 10.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 10.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 10.0 ppb
- Trace Impurities - Thallium (Tl):≤ 50.0 ppb
- Trace Impurities - Tin (Sn):≤ 50 ppb
- Trace Impurities - Titanium (Ti):≤ 100.0 ppb
- Trace Impurities - Vanadium (V):≤ 10.0 ppb
- Trace Impurities - Zinc (Zn):≤ 50 ppb
- Trace Impurities - Zirconium (Zr):≤ 10.0 ppb
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152