ContactUsLinkComponent
Gas burette, Hempel, Class A
Gas burette, Hempel, Class A
Catalog # WITG2.972.000
Supplier:  witeg Labortechnik
undefined
Gas burette, Hempel, Class A
Catalog # WITG2.972.000
Supplier:  witeg Labortechnik
Specifications
  • Capacity:
    100 ml
  • Division:
    0,2 ml
  • Pk:
    1
Specifications
About this item
Gas burette, accuracy Class A.
  • With resistant DIFFICO graduation
  • Main point ring graduation
  • Iron base