Pump tubing, Tygon® MHSL 2001
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Transparent, plasticiser-free tubing with superior pump-life.
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
Especially designed for MEK and other aggressive solvents.
Temperature resistant from −73 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88
Certifications: Meets FDA criteria for food contact