Immobilised Papain
Catalog # 786-790
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1
- Description:Papain, immobilised
Specifications
About this item
A cysteine protease enzyme (EC 3.4.22.2) immobilised on 4% agarose, cleaves Immunoglobulin G (IgG) antibody molecules in the hinge region, generating three ~50 kDa fragments; two Fab domains and a Fc domain. The papain-digested antibody is unable to promote agglutination, precipitation, opsonisation, and lysis.
- Generate Fc and Fab from IgG
- Eliminates contamination with papain enzyme
- Can be used in virtually all scenarios using free papain