Orders LinkContactUsLinkComponent
Water finding rule
Water finding rule
Water finding rule, 350 mm
Catalog # STAN14620-0
Supplier:   STANHOPE SETA
Water finding rule
Catalog # STAN14620-0
Supplier:  STANHOPE SETA
Water finding rule
Water finding rule
Water finding rule, 350 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Brass and acrylic rule, graduated and grooved on one side, to accept a thin layer of water finding paste.
  • Detects water under black oils/crude and heavy fuels
  • Used with marker pastes and dip tape to monitor depths
  • Immersion time of 20 to 60 seconds according to the product
Conforms with IP M14.
Specifications
  • Description:
    Water finding rule, 350 mm