Order Entry
Germany
ContactUsLinkComponent
Hotplates
Hotplates
  SEVEDK1042
undefined
Hotplates
  SEVEDK1042

 

  • Heating area:
    Ø 155 mm;
    Ø 185 mm
  • Heating capacity:
    1000 W;
    1500 W
  • W×D×H:
    500×290×85 mm
  • Pk:
    1
  • Plate size:
    Ø 155 mm;
    Ø 185 mm
  • Type:
    DK 1042

 

 

This hotplate features two heating elements. The exterior is white highly heat-resistant paint.

  • Stepless temperature control and pilot lights