Dissolved oxygen probes, InPro® 6800
Supplier: Mettler - Toledo
Wechseln zu:
Über diesen Artikel
The InPro O2 sensor family provides the means for reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions.
- Hygienic design, capable of withstanding CIP procedures
- Stainless steel surfaces with highly polished finish
- Steam sterilisable and autoclavable up to 140°C
- Low detection limit at high signal stability
- O-rings with FDA and USP Class VI approval
- Minimal maintenance requirement
- ISM (Intelligent Sensor Management)
3 mögliche Produktvariationen vorhanden
Product Details & Documents
The InPro O2 sensor family provides the means for reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions.
- Hygienic design, capable of withstanding CIP procedures
- Stainless steel surfaces with highly polished finish
- Steam sterilisable and autoclavable up to 140°C
- Low detection limit at high signal stability
- O-rings with FDA and USP Class VI approval
- Minimal maintenance requirement
- ISM (Intelligent Sensor Management)
Product Family Options
Produktinformationen
- TypeConcentrationMaterialConnectionØ×LForAvailabilityPrice
- METN52200966DO probe, InPro® 6800DO probe, InPro® 6800InPro® 68000 - 6 μg/l [ppb]Stainless steelVP, straight12×220 mmSpring water, fruit juices, yeast propagationAnmelden für die Verfügbarkeit1 * 1 ST
- METN52200967DO probe, InPro® 6800DO probe, InPro® 6800InPro® 68000 - 6 μg/l [ppb]Stainless steelVP, straight12×320 mmSpring water, fruit juices, yeast propagationAnmelden für die Verfügbarkeit1 * 1 ST2 946,94 €
- METN52200968DO probe, InPro® 6800DO probe, InPro® 6800InPro® 68000 - 6 μg/l [ppb]Stainless steelVP, straight12×420 mmSpring water, fruit juices, yeast propagationAnmelden für die Verfügbarkeit1 * 1 ST
Accessories
Produktinformationen
- Catalog #AvailabilityPrice
- Membrane body, T-96, 16 pieces/packMembrane body, T-96, 16 pieces/packMETN52206114Anmelden für die Verfügbarkeit1 * 16 ST - 2 338,54 €
Produktinformationen
- Catalog #AvailabilityPrice
- Membrane kit, T-96 (4 membrane bodies, 1 O-ring set, 25 ml electrolyte), for InPro 6800/6050Membrane kit, T-96 (4 membrane bodies, 1 O-ring set, 25 ml electrolyte), for InPro 6800/6050METN52200024Anmelden für die Verfügbarkeit1 * 1 ST - 646,25 €