Safety cap
Safety cap, with PTFE insert and EPDM O-ring, GL 45, 2× 1/8" tubing port, 1× 1/16" tubing port, 1× 1/4"-28 female port, PP
Catalog # JOURJR-9000-0001
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
This cap can be used for helium sparging and for delivering HPLC mobile phases. The tubing ports are made so that you push the tubing through the hole, while 1/4-28 fittings provide the best connection. Unused ports can be plugged as required.
Specifications
- Thread:GL 45
- Description:Safety cap, with PTFE insert and EPDM O-ring
- Material:PP
- Seq:01
- Connector:2× 1/8" tubing port, 1× 1/16" tubing port, 1× 1/4"-28 female port