Protein Transfer buffers
Catalog # 786-019
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 L
- Description:Western transfer buffer, Efficient™, 20X
Specifications
About this item
Efficient™ Western Transfer Buffer (20X) achieves increased protein transfer efficiency. This ready-to-use transfer buffer is prepared for optimal conductivity and efficient protein transfer, without generating excessive heat or transfer distortion.