


N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99.9% for electrophoresis
Supplier: G-Biosciences
Certificates
About this item
CAS Number:110-18-9
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335
:116.21 g/mol
Melting Pt:–64 °C
UN:2372
ADR:3,II
:GHS02
:GHS07
:GHS05
Storage Temperature:Ambient