Latex gloves, Veniclean 1340
These disposable, waterproof gloves provide limited protection against chemical splashes.
- Non sterile, powder-free, smooth finish
- High dexterity: Suitable for precision handling
- Compatible with all types of foods
- Ambidextrous
Ideal for use in laboratories, hospitals, beauty salons, care facilities and in the pharma industry.
: EN 455-1/2/3, EN ISO 374-1 Type C (KT), EN ISO 374-5 VIRUS; AQL 1,5
Food contact approved
Food contact approved
- Catalog No:
- DELPV1340/6-7
- DELPV1340/8-9
- DELPV1340/7-8
- Model:
- 1340
- 1340
- 1340
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455-1/2/3, EN ISO 374-1 Type C (KT), EN ISO 374-5 VIRUS
- EN 455-1/2/3, EN ISO 374-1 Type C (KT), EN ISO 374-5 VIRUS
- EN 455-1/2/3, EN ISO 374-1 Type C (KT), EN ISO 374-5 VIRUS
- Food contact approved:
- +
- +
- +