ContactUsLinkComponent
Wafer casettes
Wafer casettes
Catalog # PALBPCPC0101
Supplier:  Palbam
undefined
Wafer casettes
Catalog # PALBPCPC0101
Supplier:  Palbam
Specifications
  • Description:
    4 inch wafer cassette
  • Material:
    Stainless steel
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
  • Non-contaminating
  • Non-deforming
  • Fully conductive
  • Withstands temperatures up to 400 ºC
  • Designed for both manual and robotic handling