ContactUsLinkComponent
Turbidity meters, MicroTol
Turbidity meters, MicroTol
Supplier:  HF SCIENTIFIC
Certificates
Turbidity meters, MicroTol
Supplier:  HF SCIENTIFIC
Specifications
  • Catalog No:
  • 708-0022
  • 708-0023
  • 708-0024
  • 708-0025
  • 708-0026
  • 708-0027
  • Accuracy:
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • ±2% of reading or ±0,02 NTU below 40 NTU whichever is greater ±5% of reading above 40 NTU
  • Display:
  • Multi-line liquid crystal backlit display
  • Multi-line liquid crystal backlit display
  • Multi-line liquid crystal backlit display
  • Multi-line liquid crystal backlit display
  • Multi-line liquid crystal backlit display
  • Multi-line liquid crystal backlit display
  • Interfaces:
  • RS485
  • RS485
  • RS485
  • RS485
  • RS485
  • RS485
  • Operating temperature:
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • Resolution:
  • 0,0001 NTU (below 10 NTU)
  • 0,0001 NTU (below 10 NTU)
  • 0,0001 NTU (below 10 NTU)
  • 0,0001 NTU (below 10 NTU)
  • 0,0001 NTU (below 10 NTU)
  • 0,0001 NTU (below 10 NTU)
  • Response time:
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Adjustable (5 to 500 seconds) 0 - 1000 NTU
  • Sample requirements - temperature:
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • 1...50 °C (34...122 °F)
  • Power supply:
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power
  • 100 - 240 V, 47 - 63 Hz, 80 VA, AC power