Safety shower test unit
Supplier: KUNSTSTOFF GMBH GRONTZKI UND W
Certificates
About this item
Body safety showers should be tested once a month to check the functionality and the requested water flow rate of 30 L/min in an emergency case. This mobile test unit provides a very convenient and easy solution to test your body safety showers and handheld eye showers.
- Transparent PVC water reservoir on brake castors, 110 litre capacity
- Built-in suction pump (230 V) and IP 45 protected power switch
- Drain hose (2 metres) to empty the water reservoir
- Separate mounts for drain hose and power cord
- Max. diameter of the body safety shower head that fits into the big funnel: 200 mm
Centrally placed under the body safety shower the big funnel can be extended to a height of 2,30 metres, while the smaller funnel is designed to collect the water from handheld eye showers (min. flow rate: 6 L/min). The collected water amount can read on a scale at the water reservoir from 0 to 110 litres. The built-in electrical suction pump will empty the reservoir in a very short time.
W×D×H: 600×500×1730 mm
Weight: 40 kg