Orders LinkContactUsLinkComponent
Capillary columns, HP-PLOT MoleSieve
Supplier:   Agilent
Capillary columns, HP-PLOT MoleSieve
Supplier:  Agilent
Certificates
Capillary columns, HP-PLOT MoleSieve
Capillary columns, HP-PLOT MoleSieve
12 Options Available Below
Product Details & Documents
For applications requiring resolution of argon and oxygen without cryogenic cooling, select columns with thick film. The thin film configurations are ideal for many applications, including routine air monitoring and analysis. The thin film column resolves argon and oxygen at subambient temperatures.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • HEWL19091P-MS4
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    12 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 913,00 €
    913,00 €
    1 * 1 ST
       913,00 €
  • HEWL19091P-MS4E
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT Molesieve
    12 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • HEWL19091P-MS7
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    25,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 842,00 €
    842,00 €
    1 * 1 ST
       842,00 €
  • HEWL19091P-MS8
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    25,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 300,00 €
    1 300,00 €
    1 * 1 ST
       1 300,00 €
  • HEWL19091P-MS8LTM
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT Molesieve
    25,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 650,00 €
    2 650,00 €
    1 * 1 ST
       2 650,00 €
  • HEWL19095P-MS5
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    25,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 666,00 €
    666,00 €
    1 * 1 ST
       666,00 €
  • HEWL19095P-MS6
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    25,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • HEWL19095P-MS6E
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT Molesieve
    25,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €
  • HEWL19095P-MS9
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 946,00 €
    946,00 €
    1 * 1 ST
       946,00 €
  • HEWL19095P-MS0
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT MoleSieve
    50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 460,00 €
    1 460,00 €
    1 * 1 ST
       1 460,00 €
  • HEWL19095P-MS0E
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT Molesieve
    50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 640,00 €
    1 640,00 €
    1 * 1 ST
       1 640,00 €
  • HEWL19095P-MS0LTM
    Columns, HP-PLOT MoleSieve
    Columns, HP-PLOT MoleSieve
    Capillary
    HP-PLOT Molesieve
    50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 810,00 €
    2 810,00 €
    1 * 1 ST
       2 810,00 €