Disposable columns, RediSep® Rf
Catalog # TELE69-2203-380
Supplier: TELEDYNE
Specifications
- Column Format:Flash cartridge
- Particle Size:40 - 63 µm
- Pore size:60 Å
- Description:Redisep silica gel flash column
- Seq:09
- Phase:Silica
- Pk:12
Specifications
About this item
RediSep Rf disposable, bare silica columns for Flash purification of organic compounds are the benchmark for normal phase separations. This high quality media is specified for high resolution and reproducibility for everyday purifications.