Electronic crimpers and decappers
Supplier: PerkinElmer
Certificates
About this item
The electronic universal, precision control, power crimpers with adjustable settings are designed to provide multiple crimps on a single battery charge.
- Rated 1500 charge cycles
- Direct plug connection to board
- PTC's integral to battery
- Very low self-discharge
- Crimper can operate while battery is charging
Certifications: Tested to EN62133 and UN DOT