Order Entry
Germany
ContactUsLinkComponent
Cyanine 3-NHS ester chloride fluorescent labeling reagent
Cyanine 3-NHS ester chloride fluorescent labeling reagent
Catalog # 25-9004-72
Supplier:  Cytiva
CAS Number:  
Cyanine 3-NHS ester chloride fluorescent labeling reagent
Catalog # 25-9004-72
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    150 µG
  • Seq:
    1322

Specifications

About this item

 :
 :590.15 g/mol
Storage Temperature:Refrigerator
CAS Number:146368-16-3
Formula:C₃₄H₄₀ClN₃O₄