Pipette containers, SPECICON
Supplier: schuett-biotec GmbH
Jump to:
About this item
Containers are made of thick walled aluminium, anodised in gold colour.
- With sterilisation opening and bayonet closures
- Can be hermetically sealed
They can be used for the sterilisation and storage of pipettes.
2 Options Available Below
Product Details & Documents
Containers are made of thick walled aluminium, anodised in gold colour.
- With sterilisation opening and bayonet closures
- Can be hermetically sealed
They can be used for the sterilisation and storage of pipettes.
Product Family Options
Racks
Product Information
- AvailabilityPrice
- 612-1865Rack, GESTICON, stainless steel, for 4 round or rectangular pipette containers (up to 70×70 mm), Dimensions 195×229×235 mmRack, GESTICON, stainless steel, for 4 round or rectangular pipette containers (up to 70×70 mm), Dimensions 195×229×235 mmLog in for availability1 * 1 ST114,00 €
- 612-1866Rack, GESTICON, stainless steel, for 8 round or rectangular pipette containers (upto 70×70 mm), Dimensions 195×405×235 mmRack, GESTICON, stainless steel, for 8 round or rectangular pipette containers (upto 70×70 mm), Dimensions 195×405×235 mmLog in for availability1 * 1 ST146,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)SCUT3523213Log in for availability1 * 1 ST - 5,90 €