Distillation flask, STANHOPE-SETA
Supplier: STANHOPE SETA
Jump to:
About this item
For the distillation of petroleum products.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Complies with ASTM D850.
2 Options Available Below
Product Details & Documents
For the distillation of petroleum products.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Complies with ASTM D850.
Product Family Options
Product Information
- CapacityAvailabilityPrice