Manual injection valve with position sensing switches
: Agilent
The stainless steel 7725i and PEEK 9725i manual injection valves are suitable for large sample volumes, high flow rates and preparative columns sized 10 to 100 mm in diameter.
The stainless steel 7725i and PEEK 9725i manual injection valves are suitable for large sample volumes, high flow rates and preparative columns sized 10 to 100 mm in diameter.