Orders LinkContactUsLinkComponent
Capillary columns, Chromosorb 101
Capillary columns, Chromosorb 101
Supplier:   Agilent
Capillary columns, Chromosorb 101
Supplier:  Agilent
Certificates
Capillary columns, Chromosorb 101
Capillary columns, Chromosorb 101
2 Options Available Below
Product Details & Documents
Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry.
  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance
Product Family Options
Stainless steel
Product Information
  • Column Format
    Ø int.
    Length
    Ext.Ø
    Mesh size
    Availability
    Price
  • Capillary
    2,00 mm
    1,83 m
    3,18 mm
    80/100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 306,00 €
    306,00 €
    1 * 1 ST
       306,00 €
Ultimetal
Product Information
  • Column Format
    Ø int.
    Length
    Ext.Ø
    Mesh size
    Availability
    Price
  • Capillary
    2,00 mm
    1,83 m
    3,18 mm
    80/100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 306,00 €
    306,00 €
    1 * 1 ST
       306,00 €