Blood collection set, Minivette®
Catalog # SARS17.2112.050
Supplier: SARSTEDT
Specifications
- Volume:50 µl
- Description:Blood collection sets, Minivette®
- Pk:2.000
Specifications
About this item
Blood collection set for the precise dispensing of small volumes.
- Easy sample recovery
- Hygienic and secure sample transfer between patient and test equipment
Used in highly infectious nursing sectors.
Packaging: Individually wrapped.