Order Entry
Germany
ContactUsLinkComponent
Test kits, general boiler water, drop count titration, AL-94
Test kits, general boiler water, drop count titration, AL-94
Catalog # HACH222403
Supplier:  Hach
CAS Number:  
undefined
Test kits, general boiler water, drop count titration, AL-94
Catalog # HACH222403
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    General boiler water, Model AL-94 MG-L, drop count titration
  • Seq:
    01
  • Range:
    Alkalinity 5 - 400 mg/l, Chloride 5 - 400 mg/l, Hardness 0 - 2500 mg/l, Fe 0 - 4 mg/l, pH 4 - 10

Specifications

About this item

This kit is used for controlling boiler and cooling water treatment and checking corrosion using the drop count test which is a simple volumetric/ titration method with colour indication. It is an economical solution for the small boiler operator who needs to monitor alkalinity (5 to 400 mg/L), chloride (5 to 400 mg/L), hardness (0 to 2500 mg/L), iron (0 to 4 mg/L), and pH (4 to 10).

  • Simple, accurate methods
  • All-inclusive, the reagents are supplied with the kit
  • Application specific

Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.

Delivery: Supplied with ten reagents, one measuring tube, two stoppers, one colour comparator box, mixing bottle, three colour discs, instructions, and carrying case.