ContactUsLinkComponent
Contact plate, with grid, ContactClik™
Contact plate, with grid, ContactClik™
Catalog # 391-3561
Supplier:  PHOENIX BIOMEDICAL
undefined
Contact plate, with grid, ContactClik™
Catalog # 391-3561
Supplier:  PHOENIX BIOMEDICAL
Specifications
  • Ø:
    55 mm
  • Packed:
    20/sleeve, 30 sleeves/case
  • Sterility:
    Non sterile
  • Version:
    Contact plate (domed base) with grid
  • Height:
    15 mm
  • Pk:
    600
Specifications
About this item
Used for testing bioburden counts on working surfaces.
  • ContactClik™ design allows easy locking and reopening, no orientation required
  • Domed surface and media gripping tabs help the media to stick to the dish, reducing drop-off
  • Special double ring around the outside of the dish helps prevent fingers from touching the media, thus reducing the possibilities of contamination
  • Pre-treated all around the sides of the base and ready for printing a barcode, lot number or any other information as required