Orders LinkContactUsLinkComponent
Precision laboratory thermometers
Precision laboratory thermometers
Supplier:   Ludwig Schneider
Precision laboratory thermometers
Supplier:  Ludwig Schneider
Certificates
Precision laboratory thermometers
Precision laboratory thermometers
78 Options Available Below
Product Details & Documents
These total immersion thermometers are suitable for official certification.
  • Colourless prismatic capillary form
Red or blue filling, enclosed or stem type.
Ordering information: Additional thermometer models are also available (separately) with works certificate or with DAkkS calibration certificate (see bottom of ordering table).
Product Family Options
Enclosed type
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1042343
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+50 °C
    0,5 °C
    220 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
  • SCNL1042343S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+50 °C
    0,5 °C
    220 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,70 €
    53,70 €
    1 * 1 ST
       53,70 €
  • SCNL1042344S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+100 °C
    0,5 °C
    270 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,70 €
    60,70 €
    1 * 1 ST
       60,70 €
  • SCNL1042344
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+100 °C
    0,5 °C
    270 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,00 €
    41,00 €
    1 * 1 ST
       41,00 €
  • SCNL1042353
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+30 °C
    0,1 °C
    300 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,10 €
    64,10 €
    1 * 1 ST
       64,10 €
  • SCNL1042353S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+30 °C
    0,1 °C
    300 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,70 €
    86,70 €
    1 * 1 ST
       86,70 €
  • SCNL1042341
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+100 °C
    1,0 °C
    305 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
  • SCNL1042342
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    305 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,40 €
    36,40 €
    1 * 1 ST
       36,40 €
  • SCNL1042350
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+200 °C
    1,0 °C
    350 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,30 €
    43,30 €
    1 * 1 ST
       43,30 €
  • SCNL1042346S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −5...+50 °C
    0,2 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,00 €
    62,00 €
    1 * 1 ST
       62,00 €
  • SCNL1042346
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −5...+50 °C
    0,2 °C
    350 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,80 €
    41,80 €
    1 * 1 ST
       41,80 €
  • SCNL1122904S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    0...+51 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,10 €
    84,10 €
    1 * 1 ST
       84,10 €
  • SCNL1042345S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+150 °C
    0,5 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,10 €
    73,10 €
    1 * 1 ST
       73,10 €
  • SCNL1042345
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+150 °C
    0,5 °C
    350 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,60 €
    49,60 €
    1 * 1 ST
       49,60 €
  • SCNL1042604S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −38...+50 °C
    0,2 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • SCNL1042704S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −30...+30 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  • SCNL1122905S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    +47...+101 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • SCNL1122906S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    +97...+151 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • SCNL1122907S
    Enclosed type, with colourless prismatic capillary form, without official certifcation
    Enclosed type, with colourless prismatic capillary form, without official certifcation
    +147...+201 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 119,00 €
    119,00 €
    1 * 1 ST
       119,00 €
  • SCNL1042347S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −5...+100 °C
    0,2 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,30 €
    86,30 €
    1 * 1 ST
       86,30 €
  • SCNL1042347
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −5...+100 °C
    0,2 °C
    420 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,00 €
    58,00 €
    1 * 1 ST
       58,00 €
  • SCNL1042348
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    0...+50 °C
    0,1 °C
    420 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,40 €
    57,40 €
    1 * 1 ST
       57,40 €
  • SCNL1042351S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+200 °C
    0,5 °C
    450 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,10 €
    80,10 €
    1 * 1 ST
       80,10 €
  • SCNL1042351
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+200 °C
    0,5 °C
    450 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,80 €
    59,80 €
    1 * 1 ST
       59,80 €
  • SCNL1042349S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+50 °C
    0,1 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,00 €
    92,00 €
    1 * 1 ST
       92,00 €
  • SCNL1042349
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+50 °C
    0,1 °C
    420 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,50 €
    66,50 €
    1 * 1 ST
       66,50 €
  • SCNL1042703S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    0,1 °C
    550 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • SCNL1042352S
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −5...+201 °C
    0,2 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 158,00 €
    158,00 €
    1 * 1 ST
       158,00 €
  • SCNL1042354S
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    0...+100 °C
    0,1 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • SCNL1042354
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    0...+100 °C
    0,1 °C
    550 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
Stem type
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1042373
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −10...+50 °C
    0,5 °C
    220 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • SCNL1042374
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −10...+100 °C
    0,5 °C
    270 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
  • SCNL1042384
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −10...+30 °C
    0,1 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,30 €
    61,30 €
    1 * 1 ST
       61,30 €
  • SCNL1042371
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −10...+100 °C
    1,0 °C
    305 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,30 €
    26,30 €
    1 * 1 ST
       26,30 €
  • SCNL1042372
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −10...+150 °C
    1,0 °C
    305 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,70 €
    28,70 €
    1 * 1 ST
       28,70 €
  • SCNL1042375
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −10...+150 °C
    0,5 °C
    350 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,40 €
    44,40 €
    1 * 1 ST
       44,40 €
  • SCNL1042376
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −5...+50 °C
    0,2 °C
    350 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,30 €
    35,30 €
    1 * 1 ST
       35,30 €
  • SCNL1042381
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −10...+200 °C
    1,0 °C
    350 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,00 €
    41,00 €
    1 * 1 ST
       41,00 €
  • SCNL1042379
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −10...+50 °C
    0,1 °C
    420 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,50 €
    53,50 €
    1 * 1 ST
       53,50 €
  • SCNL1042377
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −5...+100 °C
    0,2 °C
    420 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,20 €
    44,20 €
    1 * 1 ST
       44,20 €
  • SCNL1042378
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    0...+50 °C
    0,1 °C
    420 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,30 €
    48,30 €
    1 * 1 ST
       48,30 €
  • SCNL1042380
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −1...+201 °C
    0,5 °C
    460 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,00 €
    59,00 €
    1 * 1 ST
       59,00 €
  • SCNL1042382
    Stem type, with yellow back round capillary form, without official certification
    Stem type, with yellow back round capillary form, without official certification
    −1...+200 °C
    0,2 °C
    610 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
With DAkkS calibration certificate
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1122904S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    0...+51 °C
    0,1 °C
    140 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 383,00 €
    383,00 €
    1 * 1 ST
       383,00 €
  • SCNL1042343S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −10...+50 °C
    0,5 °C
    220 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • SCNL1042344S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −10...+100 °C
    0,5 °C
    270 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 350,00 €
    350,00 €
    1 * 1 ST
       350,00 €
  • SCNL1042353S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −10...+30 °C
    0,1 °C
    300 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 387,00 €
    387,00 €
    1 * 1 ST
       387,00 €
  • SCNL1042345S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −10...+150 °C
    0,5 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 367,00 €
    367,00 €
    1 * 1 ST
       367,00 €
  • SCNL1042346S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −5...+50 °C
    0,2 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 352,00 €
    352,00 €
    1 * 1 ST
       352,00 €
  • SCNL1042381/04/3P
    Stem type, with yellow back round capillary form, including DAkkS-calibration certificate with 3 calibration points
    Stem type, with yellow back round capillary form, including DAkkS-calibration certificate with 3 calibration points
    −10...+200 °C
    1 °C
    350 mm
    Red
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
  • SCNL1042604S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −38…+50 °C
    0,2 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 414,00 €
    414,00 €
    1 * 1 ST
       414,00 €
  • SCNL1042704S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −30...+30 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 466,00 €
    466,00 €
    1 * 1 ST
       466,00 €
  • SCNL1122905S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    +47...+101 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 407,00 €
    407,00 €
    1 * 1 ST
       407,00 €
  • SCNL1122906S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without offical calibration
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without offical calibration
    +97...+151 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • SCNL1122907S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    +147...+201 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 408,00 €
    408,00 €
    1 * 1 ST
       408,00 €
  • SCNL1042349S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −10...+50 °C
    0,1 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 394,00 €
    394,00 €
    1 * 1 ST
       394,00 €
  • SCNL1042347S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −5...+100 °C
    0,2 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 386,00 €
    386,00 €
    1 * 1 ST
       386,00 €
  • SCNL1042351S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    −10...+200 °C
    0,5 °C
    450 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 377,00 €
    377,00 €
    1 * 1 ST
       377,00 €
  • SCNL1042703S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −38...+50 °C
    0,1 °C
    550 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 449,00 €
    449,00 €
    1 * 1 ST
       449,00 €
  • SCNL1042352S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points
    −5...+201 °C
    0,2 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
  • SCNL1042354S/04/3P
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including DAkkS-calibration certificate with 3 calibration points, without official certification
    0...+100 °C
    0,1 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 475,00 €
    475,00 €
    1 * 1 ST
       475,00 €
With Works-calibration certificate
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1122904S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    0...+51 °C
    0,1 °C
    140 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
  • SCNL1042343S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −10...+50 °C
    0,5 °C
    220 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 234,00 €
    234,00 €
    1 * 1 ST
       234,00 €
  • SCNL1042344S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −10...+100 °C
    0,5 °C
    270 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 244,00 €
    244,00 €
    1 * 1 ST
       244,00 €
  • SCNL1042353S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −10...+30 °C
    0,1 °C
    300 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €
  • SCNL1042345S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −10...+150 °C
    0,5 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • SCNL1042346S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −5...+50 °C
    0,2 °C
    350 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 246,00 €
    246,00 €
    1 * 1 ST
       246,00 €
  • SCNL1042604S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −38…+50 °C
    0,2 °C
    400 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
  • SCNL1042704S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −30...+30 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • SCNL1122905S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    +47...+101 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 284,00 €
    284,00 €
    1 * 1 ST
       284,00 €
  • SCNL1122906S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    +97...+151 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 296,00 €
    296,00 €
    1 * 1 ST
       296,00 €
  • SCNL1122907S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    +147...+201 °C
    0,1 °C
    400 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
  • SCNL1042349S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −10...+50 °C
    0,1 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 288,00 €
    288,00 €
    1 * 1 ST
       288,00 €
  • SCNL1042347S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −5...+100 °C
    0,2 °C
    420 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 264,00 €
    264,00 €
    1 * 1 ST
       264,00 €
  • SCNL1042351S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    −10...+200 °C
    0,5 °C
    450 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 271,00 €
    271,00 €
    1 * 1 ST
       271,00 €
  • SCNL1042703S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −38...+50 °C
    0,1 °C
    550 mm
    Hg, Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 349,00 €
    349,00 €
    1 * 1 ST
       349,00 €
  • SCNL1042352S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points
    −5...+201 °C
    0,2 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 379,00 €
    379,00 €
    1 * 1 ST
       379,00 €
  • SCNL1042354S/03/3P
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    Enclosed type, with colourless prismatic capillary form, including work calibration certificate with 3 calibration points, without official certification
    0...+100 °C
    0,1 °C
    550 mm
    Blue
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 370,00 €
    370,00 €
    1 * 1 ST
       370,00 €