Order Entry
Germany
ContactUsLinkComponent
Vaccum desiccator, Vacuo-Temp
Vaccum desiccator, Vacuo-Temp
Catalog # SELE4000474
Supplier:  SELECTA
undefined
Vaccum desiccator, Vacuo-Temp
Catalog # SELE4000474
Supplier:  SELECTA

Specifications

  • Weight:
    9 kg
  • W×D×H ext.:
    280×340×170 mm
  • Description:
    Heated vaccum desiccator
  • Pk:
    1

Specifications

About this item

Heated vaccum desiccator consists of an AISI 304 stainless steel outer casting and polished aluminium alloy flat surface with an effective vaccum seal.

  • Analogue vacuum gauge
  • Digital time and temperature display
  • Overheating alarm
  • Visualised parameter indicator
Delivery: Supplied with bell jar and silicon seal.