Order Entry
Germany
ContactUsLinkComponent
E.Z.N.A.® Total RNA Kit I
E.Z.N.A.® Total RNA Kit I
Supplier:  OMEGA BIO-TEK
Certificates
E.Z.N.A.® Total RNA Kit I
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Sample size:
    <1 x 10⁷ cells or 30 mg tissue
  • Binding capacity:
    100 µg
  • Yield:
    Up to 100 µg

Specifications