Nylon woven filters, CellMicroSieves™
510-0025
- Description:Disc, nylon, Ø 47 mm, pore size 50 µm
- Seq:08
- Pk:10
CellMicroSieves™ allow the rapid, yet gentle, isolation of cells, organelles and cellular debris. Unlike traditional filters, these are a precision woven matrix of pure nylon filaments that are available with pores ranging from 5 to 200 µm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. CellMicroSieves™ are biologically inert, autoclavable and can be repeatedly washed and reused. They are resistant to most acidic or basic conditions and many solvents. Extremely low protein affinity means that cells and proteins will not bind to mesh.
- Rapidly collects tissue culture cells without centrifugation
- Isolates specific cell types by allowing to pass through from a dissociated tissue sample
- Separates different sized cells from a mixture of cell types
- Concentrates and isolates cells from a bioreactor
- Acts as barrier to separate cells or tissues, while allowing free media diffusion
- Prefilters sample to prevent clogging of final filter