Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Microplate Seals
Adhesive PCR film seals, QuickSeal PCR™
Adhesive PCR film seals, QuickSeal PCR™
:
iST Scientific Ltd
Adhesive PCR film seals, QuickSeal PCR™
:
iST Scientific Ltd
Transparent, peelable film for PCR.
Non pierceable
Available sterile or non sterile
Seal integrity range: −20 to +110 °C
iST Scientific Self Adhesive Sealing Film