ContactUsLinkComponent
Hand cleanser dispenser, Eurospender, Basic
Hand cleanser dispenser, Eurospender, Basic
Catalog # BODL9770550
Supplier:  Bode
Hand cleanser dispenser, Eurospender, Basic
Catalog # BODL9770550
Supplier:  Bode
Specifications
  • Capacity:
    350/500 ml
  • Colour:
    White
  • Description:
    Hand cleanser dispenser for 350/500 ml bottles, Eurospender basic
  • Pk:
    1
Specifications
About this item
The Eurospender Basic is designed for the use of hand desinfectants, detergents and body lotion.
  • Easy pump replacement in the front
  • Easy installation, easy operation, easy cleaning
  • Dosage amounts might be adjusted from 1 to 3 ml
Delivery: Supplied with mounting device.