Order Entry
Germany
ContactUsLinkComponent
Hair thermo-hygrometer
Hair thermo-hygrometer
Catalog # 620-0621
Supplier:  MOINEAU
undefined
Hair thermo-hygrometer
Catalog # 620-0621
Supplier:  MOINEAU

Specifications

  • Range:
    –10…+55 °C; 0 - 100% RH
  • Description:
    Hair hygrometer with dial thermometer
  • Division:
    1,0 °C; 1,0% RH
  • Pk:
    1

Specifications

About this item

This hair hygrometer with analogue needle thermometer is designed for use in measuring dew point.

  • Round shape
  • Wall mounting
  • With brass case

Ø: 100 mm

Weight: 105 g