Glove liners, Puri ESD Liner Fit
Supplier: PURITECH
Certificates
About this item
These glove liners made of 100% soft nylon fabric with carbon are ideal for handling electronics.
- Hand-specific for great fit
- Excellent dexterity
- Colour-coded cuff indicates size
- Protect humans and products from static electricity
When used as inner gloves, they are also suitable for cleanroom environments.
Colour: White
Specifications
- Catalog No:
- PURTIG510ESD/XXL
- PURTIG510ESD/XL
- PURTIG510ESD/L
- PURTIG510ESD/S
- PURTIG510ESD/M_U
- Model:
- Puri ESD Liner Fit
- Puri ESD Liner Fit
- Puri ESD Liner Fit
- Puri ESD Liner Fit
- Puri ESD Liner Fit
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cleanroom class:
- 100000/ISO 8
- 100000/ISO 8
- 100000/ISO 8
- 100000/ISO 8
- 100000/ISO 8
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Anti-static:
- +
- +
- +
- +
- +